Skip to Content
Merck
All Photos(2)

Key Documents

32147

Supelco

Cloransulam-methyl

PESTANAL®, analytical standard

Synonym(s):

Methyl 3-chloro-2-{[(5-ethoxy-7-fluoro[1,2,4]triazolo[1,5-c]pyrimidine-2-yl)sulfonyl]amino}benzoate, Methyl 3-chloro-N-(5-ethoxy-7-fluoro[1,2,4]triazolo[1,5-c]pyrimidin-2-ylsulfonyl)anthranilate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13ClFN5O5S
CAS Number:
Molecular Weight:
429.81
Beilstein:
7501727
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOc1nc(F)cc2nc(nn12)S(=O)(=O)Nc3c(Cl)cccc3C(=O)OC

InChI

1S/C15H13ClFN5O5S/c1-3-27-15-18-10(17)7-11-19-14(20-22(11)15)28(24,25)21-12-8(13(23)26-2)5-4-6-9(12)16/h4-7,21H,3H2,1-2H3

InChI key

BIKACRYIQSLICJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M S Krieger et al.
Journal of chromatography. A, 897(1-2), 405-413 (2000-12-29)
Cloransulam-methyl was extracted from soil samples with supercritical CO2, subcritical water and conventional organic solvents. Supercritical CO2 was less efficient than conventional organic solvents; polarity modifiers had no impact on extraction efficiency. Extraction with supercritical CO2 exhibited a strong temperature
P Lewer et al.
Journal of agricultural and food chemistry, 48(6), 2532-2546 (2000-07-11)
Two lactating goats were given a daily oral dose of either [UL-aniline-(14)C; AN] or [triazolopyrimidine-7,9-(14)C; TP]cloransulam-methyl for 5 consecutive days. Each animal received a dietary equivalent of approximately 10 mg/kg of test material, approximately 2225 times the realistic maximum dietary
Shizuka Saito-Shida et al.
Analytical methods : advancing methods and applications, 13(7), 894-902 (2021-01-26)
A reliable multiresidue method was developed for determining multiclass acidic pesticides in cereal grains, legumes, vegetables, and fruits. The target pesticides comprise 75 compounds, including phenoxy acid, sulfonylurea, imidazoline, and triazolopyrimidine herbicides, with acidic dissociation constant (pKa) values of 1.9-5.9.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service