Skip to Content
Merck
All Photos(2)

Key Documents

43942

Supelco

Docosane

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$75.50
5 G
$295.00

$75.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$75.50
5 G
$295.00

About This Item

Linear Formula:
CH3(CH2)20CH3
CAS Number:
Molecular Weight:
310.60
Beilstein:
1702206
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$75.50


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

10.8 (vs air)

vapor pressure

<1 mmHg ( 21.1 °C)

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

369 °C (lit.)

mp

42-45 °C (lit.)
43-46 °C

density

0.778 g/mL at 25 °C (lit.)

application(s)

petroleum

format

neat

SMILES string

CCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C22H46/c1-3-5-7-9-11-13-15-17-19-21-22-20-18-16-14-12-10-8-6-4-2/h3-22H2,1-2H3

InChI key

HOWGUJZVBDQJKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Docosane can find applications as a phase change material, known as latent heat storage media, which can help in storage and conversion of solar energy.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous enhancement of latent heat and thermal conductivity of docosane-based phase change material in the presence of spongy graphene
Li.F.J, et al.
Solar Energy Materials and Solar Cells, 128, 48-51 (2014)
Auke J Kronemeijer et al.
Small (Weinheim an der Bergstrasse, Germany), 7(11), 1593-1598 (2011-05-04)
Charge transport through alkanes and para-phenylene oligomers is investigated in large-area molecular junctions. The molecules are self-assembled in a monolayer and contacted with a top electrode consisting of poly(3,4-ethylenedioxythiophene)-poly(4-styrenesulfonic acid) (PEDOT:PSS). The complete set of J(V,T) characteristics of both saturated
Carlos F Torres et al.
Biotechnology and bioengineering, 78(5), 509-516 (2002-07-13)
The effect of the water content on the lipase-catalyzed (Candida rugosa) interesterification (acidolysis) of menhaden oil with conjugated linoleic acid was studied for amounts of added water ranging from 0-4% (w/w). The rate of the acidolysis reaction increased with increasing
H Kraack et al.
The Journal of chemical physics, 121(16), 8003-8009 (2004-10-16)
The temperature dependent phase behavior of Langmuir films of n-alkanes [CH3(CH2)(n-2)CH3, denote Cn] on mercury was studied for chain lengths 19< or =n< or =22 and temperatures 15< or =T< or =44 degrees C, using surface tensiometry and surface x-ray
Shi-feng Ni et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 29(1), 54-58 (2005-02-16)
To provide the foundation for reasonable utilization by analysing the essential oils from Serissa serissoides in different seasons. Essential oils were obtained by steam distillation. The chemical components were separated and identified by gas chromatography-mass spectrometer (GC-MS). The relative content

Protocols

Separation of Decane; Dodecane; Tetradecane; Hexadecane; Octadecane; Eicosane; Docosane; Tetracosane; Hexacosane; Octacosane

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service