Skip to Content
Merck
All Photos(1)

Key Documents

31572

Supelco

Propaquizafop

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$215.00

$215.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$215.00

About This Item

Empirical Formula (Hill Notation):
C22H22ClN3O5
CAS Number:
Molecular Weight:
443.88
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$215.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

C[C@@H](Oc1ccc(Oc2cnc3cc(Cl)ccc3n2)cc1)C(=O)OCCO\N=C(\C)C

InChI

1S/C22H22ClN3O5/c1-14(2)26-29-11-10-28-22(27)15(3)30-17-5-7-18(8-6-17)31-21-13-24-20-12-16(23)4-9-19(20)25-21/h4-9,12-13,15H,10-11H2,1-3H3/t15-/m1/s1

InChI key

FROBCXTULYFHEJ-OAHLLOKOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Propaquizafop is an aryloxyphenoxypropionate herbicide primarily used for post emergence control of weeds in agricultural products.[1] Its mode of action involves inhibition of the plastid acetyl coenzyme A carboxylase (ACCase), a key enzyme in fatty acid biosynthesis.[2]

Application

Propaquizafop may be used as a reference standard for the determination of the pesticide:
  • In honeybee samples by liquid and gas chromatography coupled with tandem mass spectrometry with multiple-reaction monitoring.[3]
  • In groundwater and surface water samples by direct injection-liquid chromatography–electrospray ionization-tandem mass spectrometry with multiple-reaction monitoring.[4]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

EFFECT OF PROPAQUIZAFOP TREATMENT ON PIGMENTS CONTENT AND GROWTH OF SWEET MAIZE SEEDLINGS.
Rusjan A, et al.
Acta Scientiarum Polonorum. Technologia Alimentaria, 16(4), 227- 234 (2018)
Propaquizafop absorption, translocation, metabolism, and effect on acetyl-CoA carboxylase isoforms in chickpea (Cicer arietinum L.).
Gimenez-Espinosa R, et al.
Pesticide Biochemistry and Physiology, 65(2), 140- 150 (1999)
Multi-residue method for the determination of pesticides and pesticide metabolites in honeybees by liquid and gas chromatography coupled with tandem mass spectrometry?Honeybee poisoning incidents.
Kiljanek T, et al.
Journal of Chromatography A, 1435(4), 100-114 (2016)
A multimethod for the determination of 150 pesticide metabolites in surface water and groundwater using direct injection liquid chromatography-mass spectrometry.
Reemtsma, et al.
Journal of Chromatography A, 1271(1), 95-104 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service