Skip to Content
Merck
All Photos(1)

Key Documents

31224

Sigma-Aldrich

Chloramine T trihydrate

reag. Ph. Eur., 98.0-103.0%

Synonym(s):

N-Chloro-p-toluenesulfonamide sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$81.30
250 G
$84.30
1 KG
$235.00

$81.30


Available to ship on08 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
$81.30
250 G
$84.30
1 KG
$235.00

About This Item

Linear Formula:
CH3C6H4SO2NClNa · 3H2O
CAS Number:
Molecular Weight:
281.69
Beilstein:
3924168
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

$81.30


Available to ship on08 April 2025Details


Request a Bulk Order

Agency

USP/NF
reag. Ph. Eur.

Quality Level

Assay

98.0-103.0%

impurities

≤1.5% insoluble in ethanol

pH

8-10 (20 °C, 5%)

mp

167-170 °C (lit.)

SMILES string

O.O.O.Cc1ccc(cc1)S(=O)(=O)N([Na])Cl

InChI

1S/C7H7ClNO2S.Na.3H2O/c1-6-2-4-7(5-3-6)12(10,11)9-8;;;;/h2-5H,1H3;;3*1H2/q-1;+1;;;

InChI key

NZYOAGBNMCVQIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloramine-T trihydrate is a widely used oxidizing agent in organic synthesis. It acts as a source of halonium cation and nitrogen anion and thus acts as a base and nucleophile. Due to its reactivity with a wide range of functional groups, it is involved in different molecular transformations. It is effective in acidic, neutral, and basic conditions[1].

Application

Chloramine-T trihydrate can be used as:
  • A reagent for chlorination in drinking water treatment[1].
  • A dehydrogenating agent for the oxidative dehydrogenation of aromatic aldehyde phenylhydrazones to synthesize nitrile imines[2].
  • The nitrene source for the aziridination of alkenes and the amidation of cyclic ethers[3].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

377.6 °F - closed cup

Flash Point(C)

192 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chloramine-T (N-chloro-p-toluenesulfonamide sodium salt), a versatile reagent in organic synthesis and analytical chemistry: An up to date review
Nayak Y, et al.
Journal of Saudi Chemical Society, 101416-101416 (2022)
Nitrene transfer reactions catalysed by copper (I) complexes in ionic liquid using chloramine-T
Cano I, et al.
Dalton Transactions (2007)
Crystal and molecular structure of chloramine-T trihydrate. Absence of a sodium-nitrogen interaction in the oxidant. N-chloro-N-sodiotoluene-p-sulfonamide.
Olmstead MM and Power PP.
Inorganic Chemistry, 25(22), 4057-4058 (1986)
Synthesis of 3, 4-diaryl-1-phenyl-4, 5-dihydro-1H-pyrazole-5-carbonitriles via 1, 3-dipolar cycloaddition reactions
Prabhashankar J, et al.
Turkish Journal of Chemistry, 37, 853-857 (2013)
Chloramine T and related N-halogeno-N-metallo reagents.
Campbell MM and Johnson G.
Chemical Reviews, 78(1), 65-87 (1978)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service