Skip to Content
Merck
All Photos(1)

Key Documents

03891

Supelco

Ehrlich′s solution

2% 4-(n,n-Dimethylamino)-benzaldehyde in 22% hydrochloric acid

Synonym(s):

4-(Dimethylamino)benzaldehyde – hydrochloric acid solution

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$134.00

$134.00


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 ML
$134.00

About This Item

CAS Number:
Beilstein:
4132845
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.25

$134.00


Available to ship on07 April 2025Details


Request a Bulk Order

Product Name

Ehrlich′s solution,

form

liquid

Quality Level

density

1.10 g/mL at 20 °C

application(s)

food and beverages

SMILES string

CN(C)c1ccc(C=O)cc1

InChI

1S/C9H11NO/c1-10(2)9-5-3-8(7-11)4-6-9/h3-7H,1-2H3

InChI key

BGNGWHSBYQYVRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

reagent for urobilinogen

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service