Skip to Content
Merck
All Photos(3)

Key Documents

296104

Sigma-Aldrich

1,2,4-Trichlorobenzene

anhydrous, ≥99%

Synonym(s):

1,2,4-TCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl3
CAS Number:
Molecular Weight:
181.45
Beilstein:
956819
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
≥99%
bp:
214 °C (lit.)
vapor pressure:
1 mmHg ( 40 °C)
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor density

>6 (vs air)

vapor pressure

1 mmHg ( 40 °C)

Assay

≥99%

form

solid or liquid

autoignition temp.

1060 °F

expl. lim.

6.6 %, 150 °F

impurities

<0.002% water
<0.005% water (100 mL pkg)

refractive index

n20/D 1.571 (lit.)

bp

214 °C (lit.)

mp

16 °C (lit.)

solubility

water: insoluble

density

1.454 g/mL at 25 °C (lit.)

SMILES string

Clc1ccc(Cl)c(Cl)c1

InChI

1S/C6H3Cl3/c7-4-1-2-5(8)6(9)3-4/h1-3H

InChI key

PBKONEOXTCPAFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2,4-Trichlorobenzene is a highly chlorinated aromatic solvent. It is mainly used as intermediate in dyes, pesticides and also as a lubricant.[1]

Application

1,2,4-Trichlorobenzene may be used as a solvent to prepare:
  • dimethylketene β-lactone dimer from tetramethyl-1,3-cyclobutanedione[2]
  • salicyl-o-toluide by the reaction of phenyl salicylate and o-toluidine[3]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dimethylketene ?-lactone dimer
Robert H, et al.
Organic Syntheses, 48, 72-72 (1968)
Biomineralisation of 1,2,4-trichlorobenzene in soils by an adapted microbial population.
Schroll R, et al.
Environmental Pollution (Barking, Essex : 1987), 127(3), 395-401 (2004)
Salicyl-o-toluide
Allen CFH and VanAllan J
Organic Syntheses, 26, 92-92 (1946)
Synthesis and characterization of several Ni/NiAl2O4 catalysts active for the 1, 2, 4-trichlorobenzene hydrodechlorination.
Cesteros Y, et al.
Applied Catalysis. B, Environmental, 25(4), 213-227 (2000)
The effect of trichlorobenzene solvent geometry on the morphologies of C60 nano/microcrystals produced from solution.
Liu DD, et al.
Materials Research Bulletin, 73, 65-69 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service