Skip to Content
Merck
All Photos(1)

Key Documents

8.21152

Sigma-Aldrich

1,2,4-Trichlorobenzene

for synthesis

Synonym(s):

1,2,4-Trichlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3-1,2,4-(Cl)3
CAS Number:
Molecular Weight:
181.45
MDL number:
UNSPSC Code:
12352101
EC Index Number:
204-428-0
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
213.5 °C/1013 hPa
vapor pressure:
1.3 hPa ( 20 °C)
Pricing and availability is not currently available.

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

1.3 hPa ( 20 °C)

Assay

≥98% (GC)

form

liquid

autoignition temp.

571 °C

potency

550 mg/kg LD50, oral (Rat)
6139 mg/kg LD50, skin (Rat)

expl. lim.

2.5-6.6 % (v/v)

dilution

(for synthesis)

bp

213.5 °C/1013 hPa

mp

17 °C

transition temp

flash point 99 °C

density

1.45 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Clc1c(ccc(c1)Cl)Cl

InChI

1S/C6H3Cl3/c7-4-1-2-5(8)6(9)3-4/h1-3H

InChI key

PBKONEOXTCPAFI-UHFFFAOYSA-N

Related Categories

Application


  • Toxicity of xenobiotics during sulfate, iron, and nitrate reduction in primary sewage sludge suspensions.: This study discusses the toxicological impacts of various xenobiotics, including 1,2,4-trichlorobenzene, in the context of environmental chemistry, highlighting the reduction processes in sewage sludge. This is relevant for those investigating the environmental fate and transport of chemical pollutants (Elsgaard et al., 2010).

  • Multigeneration study of 1,2,4-trichlorobenzene in rats.: This research examines the effects of 1,2,4-trichlorobenzene over multiple generations in rats, offering insights into the compound′s long-term biological impact, crucial for environmental health perspectives and risk assessment studies (Robinson et al., 1981).


Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.453 - 1.456
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service