Skip to Content
Merck
All Photos(3)

Key Documents

290009

Sigma-Aldrich

Potassium benzoate

ReagentPlus®, 99%

Synonym(s):

Benzoic acid potassium salt

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$106.00
1 KG
$182.00

$106.00


Available to ship on10 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
$106.00
1 KG
$182.00

About This Item

Linear Formula:
C6H5COOK
CAS Number:
Molecular Weight:
160.21
Beilstein:
3719165
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent

$106.00


Available to ship on10 April 2025Details


Request a Bulk Order

grade

reagent

Quality Level

product line

ReagentPlus®

Assay

99%

functional group

phenyl

SMILES string

[K+].[O-]C(=O)c1ccccc1

InChI

1S/C7H6O2.K/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);/q;+1/p-1

InChI key

XAEFZNCEHLXOMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microphase Separation with Small Amphiphilic Molecules: Crystal Structure of Preservatives Sodium Benzoate (E 211) and Potassium Benzoate (E 212).
Butterhof C, et al.
Zeitschrift fur Anorganische und Allgemeine Chemie, 639(15), 2816-2821 (2013)
N Zengin et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 49(4), 763-769 (2010-12-07)
In this study, the genotoxic effects of sodium benzoate (SB) and potassium benzoate (PB) were investigated in cultured human peripheral lymphocytes using chromosomal aberrations (CA), sister chromatid exchange (SCE), and micronuclei (MN). The level of nuclear DNA damage of SB
Polylactones, 19. Anionic polymerization of L-lactide in solution.
Kricheldorf HR and Kreiser-Saunders I.
Makromol. Chem., 191(5), 1057-1066 (1990)
Chunhua Liu et al.
European journal of pharmacology, 700(1-3), 118-126 (2013-01-12)
Leonurine possesses cardioprotective effects in myocardial ischemia due to its anti-apoptotic properties. However, the process to isolate and purify leonurine is difficult, because of its low content in the Herb Leonuri and its impurity. Moreover, the high dosage used indicates
Renbo Wei et al.
Macromolecular rapid communications, 34(4), 330-334 (2013-01-03)
A new approach is developed to fabricate highly oriented mono-domain LCE nano/microstructures through micro-molding in capillaries. Gratings and microwires as two typical examples are fabricated and characterized by polarizing optical microscopy, optical microscopy, and scanning electron microscopy. The gratings with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service