28621-U
Hamilton® 1001LTN syringe
1000LTN, volume 1 mL, needle size 23 ga (cone tip, side port), needle L 56 mm (2.2 in.)
Synonym(s):
1001LTN High-Pressure Syringe
About This Item
Recommended Products
material
cone tip, side port stainless steel (point style 5)
needle size
23 ga (cone tip, side port)
description
gastight
feature
GASTIGHT® Syringe, Fixed Needle
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 203082
needle L
56 mm (2.2 in.)
volume
1 mL
compatibility
for use with CTC COMBI PAL (luer tip needle)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Rapid extraction of BTEX in water and milk samples based on functionalized multi-walled carbon nanotubes by dispersive homogenized-micro-solid phase extraction.: This study utilizes the Hamilton® 1001LTN syringe for precise sample injection in the extraction of BTEX (benzene, toluene, ethylbenzene, and xylene) from water and milk samples. The method demonstrates the syringe′s effectiveness in achieving high extraction efficiency and accurate analytical results (Teimoori et al., 2023).
- Bioprospecting of Essential Oil-Bearing Plants: Rapid Screening of Volatile Organic Compounds Using Headspace Bubble-in-Drop Single-Drop Microextraction for Gas Chromatography Analysis.: This research employs the Hamilton® 1001LTN syringe in a novel headspace microextraction technique for the analysis of volatile organic compounds in essential oils. The syringe′s precision facilitates the efficient screening of plant volatiles, enhancing the study of essential oil profiles (Letseka et al., 2022).
- Validation of an Analytical Method for Determination of 13 priority polycyclic aromatic hydrocarbons in mineral water using dispersive liquid-liquid microextraction and GC-MS.: This study validates an analytical method incorporating the Hamilton® 1001LTN syringe for the precise injection of mineral water samples. The syringe′s accuracy is pivotal in detecting low levels of polycyclic aromatic hydrocarbons, contributing to reliable environmental monitoring (Sadeghi et al., 2016).
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service