Skip to Content
Merck
All Photos(1)

Documents

25539

Supelco

Hexyl acetate

analytical standard

Synonym(s):

Capryl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COO(CH2)5CH3
CAS Number:
Molecular Weight:
144.21
Beilstein:
1747138
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.7% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.409 (lit.)
n20/D 1.409

bp

168-170 °C (lit.)

mp

−80 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCOC(C)=O

InChI

1S/C8H16O2/c1-3-4-5-6-7-10-8(2)9/h3-7H2,1-2H3

InChI key

AOGQPLXWSUTHQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexyl acetate is a short-chain ester and an aromatic compound with a green note flavor, widely used in food industries.

Application

Hexyl acetate may be used as a volatile aroma reference standard for the analysis of the analyte in apples using solid-phase microextraction (SPME), time-compressed chromatography (TCC), and time-of-flight mass spectrometry (TOFMS) methods.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid analysis of volatile flavor compounds in apple fruit using SPME and GC/time-of-flight mass spectrometry.
Song J, et al.
Journal of Agricultural and Food Chemistry, 45(5), 1801-1807 (1997)
Optimized synthesis of lipase-catalyzed hexyl acetate in n-hexane by response surface methodology.
Shieh CJ and Chang SW
Journal of Agricultural and Food Chemistry, 49(3), 1203-1207 (2001)
Zhilin Zhang et al.
Journal of insect science (Online), 14 (2014-12-21)
The morphological structure of the metathoracic scent glands (MTGs) in Adelphocoris suturalis was observed by utilizing scanning electron microscope (SEM). Also, the secretions of MTGs in male and female were analyzed by using gas chromatography-mass spectrometry (GC-MS). The result showed
Rachel S Leisso et al.
Physiologia plantarum, 153(2), 204-220 (2014-06-20)
'Soggy breakdown' (SB) is an internal flesh disorder of 'Honeycrisp' apple (Malus × domestica Borkh.) fruit that occurs during low temperature storage. The disorder is a chilling injury (CI) in which visible symptoms typically appear after several weeks of storage
Zuobing Xiao et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 978-979, 122-130 (2014-12-30)
A simple, rapid and solvent-free multi-residue method has been developed and applied to confirm and quantify a series of volatile compounds in five cherry wines by gas chromatography coupled with mass spectrometry (GC-MS). Four parameters (e.g., coating material of fiber

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service