Skip to Content
Merck
All Photos(4)

Key Documents

252751

Sigma-Aldrich

3-Ethoxy-4-methoxybenzaldehyde

99%

Synonym(s):

3-Ethoxy-p-anisaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$91.50

$91.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$91.50

About This Item

Linear Formula:
C2H5OC6H3(OCH3)CHO
CAS Number:
Molecular Weight:
180.20
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$91.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

powder, crystals or chunks

mp

51-53 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCOc1cc(C=O)ccc1OC

InChI

1S/C10H12O3/c1-3-13-10-6-8(7-11)4-5-9(10)12-2/h4-7H,3H2,1-2H3

InChI key

VAMZHXWLGRQSJS-UHFFFAOYSA-N

General description

3-Ethoxy-4-methoxybenzaldehyde is a derivative of vanillin.

Application

3-Ethoxy-4-methoxybenzaldehyde may be used to synthesize Compound of Designation red 10 binding (CDr10b) to investigate its role in neuroinflammatory diseases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service