Skip to Content
Merck
All Photos(2)

Key Documents

173606

Sigma-Aldrich

4-Ethoxybenzaldehyde

99%

Synonym(s):

p-Ethoxybenzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$68.50
100 G
$174.00

$68.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$68.50
100 G
$174.00

About This Item

Linear Formula:
C2H5OC6H4CHO
CAS Number:
Molecular Weight:
150.17
Beilstein:
386863
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$68.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.559 (lit.)

bp

255 °C (lit.)

mp

13-14 °C (lit.)

density

1.08 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)c1ccc(OCC)cc1

InChI

1S/C9H10O2/c1-2-11-9-5-3-8(7-10)4-6-9/h3-7H,2H2,1H3

InChI key

JRHHJNMASOIRDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Ethoxybenzaldehyde was used as an anti-inflammatory and prostaglandin E2 suppressor in skin lightening agents[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth T T Makino et al.
Journal of drugs in dermatology : JDD, 12(3), s16-s20 (2013-04-03)
Skin lightening preparations are used by people all over the world for a diverse range of dermatologic indications. Hydroquinone (HQ) is the gold standard and remains the only prescription product available in the United States for the treatment of generalized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service