24137-M
2-Propanol
≥99.5% (GC), puriss., meets analytical specification of Ph. Eur., BP, USP
Synonym(s):
sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol
About This Item
44 mmHg ( 25 °C)
Recommended Products
Product Name
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
vapor density
2.1 (vs air)
Quality Level
vapor pressure
33 mmHg ( 20 °C)
44 mmHg ( 25 °C)
grade
puriss.
Assay
≥99.5% (GC)
autoignition temp.
750 °F
quality
meets analytical specification of Ph. Eur., BP, USP
expl. lim.
2.0-12.7 %, 93 °C
impurities
acidity or alkalinity, complies
benzene and related subst., complies
peroxides, complies
residual solvents, complies
≤0.0001% benzene
≤0.0002% heavy metals (as Pb)
≤0.002% free acid (as C2H5COOH)
≤0.002% non-volatile matter
≤0.05% aldehydes and ketones (as CH3COCH3)
≤0.1% methanol (GC)
≤0.2% water (Karl Fischer)
refractive index
n20/D 1.377 (lit.)
n20/D 1.377-1.378
bp
82 °C (lit.)
mp
−89.5 °C (lit.)
density
0.785 g/mL at 25 °C (lit.)
cation traces
Fe: ≤0.0001%
absorption
in accordance
suitability
complies for appearance of solution
complies for identity
SMILES string
CC(C)O
InChI
1S/C3H8O/c1-3(2)4/h3-4H,1-2H3
InChI key
KFZMGEQAYNKOFK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Component in a binary mixture for the determination of vapor-liquid equilibrium data.
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
53.6 °F - closed cup
Flash Point(C)
12.0 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service