Skip to Content
Merck
All Photos(5)

Key Documents

8.18766

Supelco

2-Propanol

≥99.5% (GC), EMPLURA®

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$36.90
2.5 L
$76.80
10 L
$270.00
25 L
$456.00
180 L
$2,810.00

$36.90


Estimated to ship on05 May 2025FromTruganina


Request a Bulk Order

Select a Size

Change View
1 L
$36.90
2.5 L
$76.80
10 L
$270.00
25 L
$456.00
180 L
$2,810.00

About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-661-7
NACRES:
NB.21
bp:
82 °C (lit.)
vapor pressure:
33 mmHg ( 20 °C)
43 hPa ( 20 °C)
44 mmHg ( 25 °C)

$36.90


Estimated to ship on05 May 2025FromTruganina


Request a Bulk Order

Product Name

2-Propanol, EMPLURA®

vapor density

2.1 (vs air)

Quality Level

vapor pressure

33 mmHg ( 20 °C)
43 hPa ( 20 °C)
44 mmHg ( 25 °C)

product line

EMPLURA®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

750 °F

potency

5045 mg/kg LD50, oral (Rat)
12800 mg/kg LD50, skin (Rabbit)

expl. lim.

2.0-12.7 %, 93 °C

dilution

(low-cost alternative, for basic applications)

impurities

≤0.2% Water

evapn. residue

≤0.0020%

refractive index

n20/D 1.377 (lit.)

pH

7 (20 °C)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

transition temp

flash point 12 °C

density

0.785 g/mL at 25 °C (lit.)

storage temp.

2-25°C

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application


  • Asymmetric Catalysis by Architectural and Functional Molecular Engineering: Practical Chemo- and Stereoselective Hydrogenation of Ketones.: The study showcases the role of 2-propanol in asymmetric catalysis, providing an efficient method for the stereoselective hydrogenation of ketones, which is vital in pharmaceutical synthesis (Noyori and Ohkuma, 2001).

  • Electrospray ionization mass spectrometry of synthetic oligonucleotides using 2-propanol and spermidine.: This research examines the use of 2-propanol in electrospray ionization mass spectrometry, enhancing the detection and analysis of synthetic oligonucleotides (De Bellis et al., 2000).

  • Isolation, identification, and chemical synthesis of 8 alpha,25-dihydroxy-9,10-seco-4,6,10(19)-cholestatrien-3-one. A new metabolite of 25-hydroxyvitamin D3 produced by mouse myeloid leukemia cells (M1).: This study details the use of 2-propanol in the chemical synthesis of a new vitamin D3 metabolite, which is significant for understanding vitamin D metabolism in leukemia cells (Yamada et al., 1987).

Analysis Note

Purity (GC): ≥ 99.5 %
Identity (IR): conforms
Density (d 20 °C/ 4 °C): 0.784 - 0.785
Evaporation residue: ≤ 0.0020 %
Water: ≤ 0.20 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service