Skip to Content
Merck
All Photos(1)

Key Documents

20742

Syringe Cleaning Solution

size 70 mL

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$52.50

$52.50


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$52.50

About This Item

UNSPSC Code:
41122001
eCl@ss:
32070500

$52.50


Estimated to ship on30 May 2025


Request a Bulk Order

packaging

pkg of 70 mL

manufacturer/tradename

Hamilton 18310

size

70 mL

Other Notes

Biodegradable cleaning concentrate helps to eliminate hard to remove residues.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Syringes are precision instruments and need care. Learn to correctly inspect, use, unclog, handle, clean and store them to ensure good accuracy, reproducibility, and lifetime.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service