Key Documents
20714
Hamilton® GASTIGHT® syringe, 1700 series, luer tip
1750LT, volume 0.5 mL, without needle, Luer Tip, needle size (not included)
Select a Size
1 EA
$115.00
$115.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$115.00
About This Item
UNSPSC Code:
41122001
eCl@ss:
32070500
$115.00
Check Cart for Availability
Request a Bulk Order
material
glass body
needle size
(not included)
feature
without needle Luer Tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81201
needle L
, n/a
volume
0.5 mL
Related Categories
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. What is the meaning of GASTIGHT®? Are GASTIGHT® syringes suitable for use with liquid samples?
    1 answer
    1. The term GASTIGHT® does not prohibit the use of a GASTIGHT® syringe with liquids. GASTIGHT® denotes that the syringe plunger features a Teflon tip, which prevents the sample from creeping up around the barrel, particularly crucial when working with solvents.
      Helpful?