Skip to Content
Merck
All Photos(1)

Documents

18804

Supelco

Crocetin dialdehyde

analytical standard

Synonym(s):

(all-E)-2,6,11,15-Tetramethyl-2,4,6,8,10,12,14-hexadecaheptaenedial, 8,8′-Diapo-ψ,ψ-carotenedial, Crocetindial

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O2
CAS Number:
Molecular Weight:
296.40
Beilstein:
1713035
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[H]C(/C(C)=C/C=C/C(C)=C/C=C/C=C(C)/C=C/C=C(C)/C([H])=O)=O

InChI

1S/C20H24O2/c1-17(11-7-13-19(3)15-21)9-5-6-10-18(2)12-8-14-20(4)16-22/h5-16H,1-4H3/b6-5+,11-7+,12-8+,17-9+,18-10+,19-13+,20-14+

InChI key

YHCIKUXPWFLCFN-QHUUTLAPSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhichao Xu et al.
BMC biology, 18(1), 63-63 (2020-06-20)
Plants have evolved a panoply of specialized metabolites that increase their environmental fitness. Two examples are caffeine, a purine psychotropic alkaloid, and crocins, a group of glycosylated apocarotenoid pigments. Both classes of compounds are found in a handful of distantly
Keizo Umegaki et al.
Journal of oleo science, 68(10), 995-1002 (2019-10-05)
Coleus forskohlii extract (CFE), a popular weight-loss herbal product, induces hepatic cytochrome P450 (CYP) and fatty liver in mice; however, its main bioactive ingredient, forskolin, does not show such effects. To ensure the safety of CFE as a dietary supplement

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service