Skip to Content
Merck
All Photos(1)

Key Documents

SML3255

Sigma-Aldrich

Crocetin

≥98% (HPLC)

Synonym(s):

α-Crocetin, 2,6,11,15-tetramethyl-2E,4E,6E,8E,10E,12E,14E-hexadecaheptaenedioic acid, NSC 407300, Trans-crocetin, Transcrocetin

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$14.60
25 MG
$72.80

$14.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$14.60
25 MG
$72.80

About This Item

Empirical Formula (Hill Notation):
C20H24O4
CAS Number:
Molecular Weight:
328.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$14.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

faint brown to dark brown
deep red-brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

−70°C

SMILES string

OC(=O)\C(=C\C=C\C(=C\C=C\C=C(/C)\C=C\C=C(/C)\C(=O)O)\C)\C

InChI

1S/C20H24O4/c1-15(11-7-13-17(3)19(21)22)9-5-6-10-16(2)12-8-14-18(4)20(23)24/h5-14H,1-4H3,(H,21,22)(H,23,24)/b6-5+,11-7+,12-8+,15-9+,16-10+,17-13+,18-14+

InChI key

PANKHBYNKQNAHN-MQQNZMFNSA-N

Biochem/physiol Actions

Apocarotenoid dicarboxylic acid shown to exhibit antioxidant, antiproliferative, anti-inflammatory, cardioprotective, and antinociceptive activity.
Crocetin is a natural apocarotenoid dicarboxylic acid, found naturally in saffron crocus and gardenia flowers, that has been shown to exhibit antioxidant, antiproliferative, anti-inflammatory, cardioprotective, neuroprotective, and antinociceptive activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eiji Inoue et al.
Journal of natural medicines, 72(1), 274-279 (2017-11-18)
Saffron, the stigma of Crocus sativus Linné (Iridaceae family), has been known to inhibit aggregation of β-amyloid, a nerve tissue protein. α-Synuclein (αS) is a 140-amino acid protein found abundantly in various regions of the brain. Its abnormal aggregation and
Kyriaki Hatziagapiou et al.
Current neuropharmacology, 17(4), 377-402 (2018-03-23)
Reactive oxygen species and reactive nitrogen species, which are collectively called reactive oxygen-nitrogen species, are the inevitable by-products of cellular metabolic redox reactions, such as oxidative phosphorylation in the mitochondrial respiratory chain, phagocytosis, reactions of biotransformation of exogenous and endogenous
Maliheh Moradzadeh et al.
Journal of cellular physiology, 233(3), 2170-2182 (2017-04-14)
Natural products have gained a wide popularity as chemopreventive and anti-cancer agents owing to their multi-mechanistic mode of action, availability and synergism with several conventional chemotherapeutic agents. Crocetin is a carotenoid compound isolated from the stigma of Crocus sativus L.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service