Skip to Content
Merck
All Photos(1)

Key Documents

14400

Sigma-Aldrich

Biotin

≥99.0% (T)

Synonym(s):

D-Biotin, Bios II, Coenzyme R, Vitamin B7, Vitamin H

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$50.40
1 G
$190.00
5 G
$740.00

$50.40


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 MG
$50.40
1 G
$190.00
5 G
$740.00

About This Item

Empirical Formula (Hill Notation):
C10H16N2O3S
CAS Number:
Molecular Weight:
244.31
Beilstein:
86838
EC Number:
MDL number:
UNSPSC Code:
12352201
eCl@ss:
34058011
PubChem Substance ID:
NACRES:
NA.21

$50.40


Available to ship on07 April 2025Details


Request a Bulk Order

biological source

synthetic

Quality Level

Assay

≥99.0% (T)

form

powder

optical activity

[α]20/D +91±2°, c = 1% in 0.1 M NaOH

ign. residue

≤0.1% (as SO4)

loss

≤0.5% loss on drying

color

colorless to white

mp

229-234 °C
231-233 °C (lit.)

solubility

alcohol: soluble 80mg/100 ml at 25 °C
water: soluble 22mg/100 ml at 25 °C
0.1 M NaOH: 10 mg/mL at 25 °C, clear, colorless
H2O: very soluble
organic solvents: insoluble

cation traces

Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤10 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤500 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

detection

SMILES string

[H][C@]12CS[C@@H](CCCCC(O)=O)[C@@]1([H])NC(=O)N2

InChI

1S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7-,9-/m0/s1

InChI key

YBJHBAHKTGYVGT-ZKWXMUAHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Biotin is readily available in several foods, like whole grains, nuts, legumes, unpolished rice and egg yolk. The normal gut flora also produces this vitamin.[1]

Application

Biotin has been used for testing the functionality of the Avidin-displaying phages.[2] The product has also been used for labeling the swine anti-rabbit IgG antibody and peroxidase.[3]

Biochem/physiol Actions

Biotin acts as an important cofactor for carboxylase enzymes in various metabolic pathways. Biotin favors keratin synthesis and hence is used as a supplement for hair and nail growth. Deficiency of biotin shows several symptoms such as alopecia, conjunctivitis, seborrheic dermatitis, depression, lethargy, hypotonia and seizures.[1]
Biotin also referred to as vitamin H or vitamin B7 is a water-soluble B-vitamin that induces two to three fold increase in guanylate cyclase activity in rat liver, kidney, colon, cerebellum, and heart.[4] It also stimulates the sperm motility and prolongs the survival of cryopreserved human spermatozoa.[5]

Other Notes

Reviews[6][7]; Avidin-Biotin technology;

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Wilchek and E.A. Bayer, eds.
Methods of Enzymatic Analysis, 184 (1990)
H. Katsuki et al.
Antibiotics, Vitamins and Hormones (1977)
Biotin.
P N Murthy et al.
Progress in food & nutrition science, 2(9), 405-455 (1977-01-01)
Guruprasad Kalthur et al.
Journal of assisted reproduction and genetics, 29(7), 631-635 (2012-04-25)
To study the effect of supplementing biotin to sperm preparation medium on the motility of frozen-thawed spermatozoa. Semen samples of men attending the University infertility clinic (n = 105) were cryopreserved using glycerol-egg yolk-citrate buffered cryoprotective medium in liquid nitrogen. After a
Tiina A Riihimäki et al.
BMC biotechnology, 11, 64-64 (2011-06-11)
Engineered proteins, with non-immunoglobulin scaffolds, have become an important alternative to antibodies in many biotechnical and therapeutic applications. When compared to antibodies, tailored proteins may provide advantageous properties such as a smaller size or a more stable structure. Avidin is

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service