Skip to Content
Merck
All Photos(1)

Documents

914479

Sigma-Aldrich

Biotin-O2Oc-O2Oc-OH

≥95%

Synonym(s):

10,19-Dioxo-23-(2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-3,6,12,15-tetraoxa-9,18-diazatricosanoic acid, 17-Biotinamido- 10-oxo-3,6,12,15-tetraoxa-9-azaheptadecan-1-oic acid, 8- Biotinylamido-3,6-dioxaoctanoic acid dimer, Acid-terminated biotin linker, Biotin-AEEA-AEEA-OH, Biotin-AEEAc-AEEAc-OH, Biotin-Ado-Ado-OH, Biotinylation reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H38N4O9S
Molecular Weight:
534.62
UNSPSC Code:
12352200

Assay

≥95%

form

powder

storage temp.

−20°C

Application

Biotin-O2Oc-O2Oc-OH is a versatile biotinylated linker that can be incorporated into chemical tools via its terminal amino group. Labeling materials or proteins with biotin provides a means to enrich and capture targets from biological systems.

Automate your Biotin tagging with Synple Automated Synthesis Platform (SYNPLE-SC002)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service