Skip to Content
Merck
All Photos(1)

Key Documents

12801

Sigma-Aldrich

Ammonium tetrachlorocuprate(II) dihydrate

purum, ≥98%

Synonym(s):

Diammonium tetrachlorocuprate(II) dihydrate, Ammonium chlorocuprate(II), Ammonium copper chloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$104.00
6 X 1 KG
$525.00

$104.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
$104.00
6 X 1 KG
$525.00

About This Item

Linear Formula:
(NH4)2CuCl4 · 2H2O
CAS Number:
Molecular Weight:
277.47
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$104.00


Check Cart for Availability

Request a Bulk Order

grade

purum

Quality Level

Assay

≥98%

form

solid

reaction suitability

core: copper
reagent type: catalyst

anion traces

sulfate (SO42-): ≤100 mg/kg

cation traces

Fe: ≤50 mg/kg

SMILES string

[NH4+].[NH4+].O.O.Cl[Cu--](Cl)(Cl)Cl

InChI

1S/4ClH.Cu.2H3N.2H2O/h4*1H;;2*1H3;2*1H2/q;;;;+2;;;;/p-2

InChI key

LLMDTWKBBWCWNN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Detection of iron in steel

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service