Skip to Content
Merck
All Photos(1)

Key Documents

00634

Sigma-Aldrich

Ammonium tetrachlorocuprate(II) dihydrate

puriss., ≥99.0% (AT)

Synonym(s):

Ammonium chlorocuprate(II), Ammonium copper chloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$87.10
1 KG
$208.00

$87.10


Available to ship on08 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
$87.10
1 KG
$208.00

About This Item

Linear Formula:
(NH4)2CuCl4 · 2H2O
CAS Number:
Molecular Weight:
277.47
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

$87.10


Available to ship on08 April 2025Details


Request a Bulk Order

grade

puriss.

Quality Level

Assay

≥99.0% (AT)

reaction suitability

core: copper
reagent type: catalyst

impurities

≤0.005% SO4
≤0.05% free acid (as HCl)

cation traces

Fe: ≤20 mg/kg

SMILES string

[NH4+].[NH4+].O.O.Cl[Cu--](Cl)(Cl)Cl

InChI

1S/4ClH.Cu.2H3N.2H2O/h4*1H;;2*1H3;2*1H2/q;;;;+2;;;;/p-2

InChI key

LLMDTWKBBWCWNN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium tetrachlorocuprate(II) dihydrate has been used in the preparation of starch cuprate via microwave-assisted cupration of potato starch.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterisation of starch cuprate.
Staroszczyk H.
Food Chemistry, 129(3), 1217-1223 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service