Skip to Content
Merck
All Photos(1)

Key Documents

09929

Sigma-Aldrich

Leit-C

for microscopy

Synonym(s):

Conduct-C

Sign Into View Organizational & Contract Pricing

Select a Size

30 G
$117.00

$117.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
30 G
$117.00

About This Item

MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

$117.00


Check Cart for Availability

Request a Bulk Order

grade

for microscopy

Quality Level

impurities

xylene (mixture of isomers)

suitability

suitable for microscopy

Application

suspension of minute, conductive carbon particles for the production of conductivity joints

Signal Word

Danger

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Central nervous system,Liver,Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

<32.0 °F - closed cup

Flash Point(C)

< 0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service