Skip to Content
Merck
All Photos(1)

Key Documents

06441

Supelco

Amberlite IRA-96 Ion Exchange Resin

free base, 16-50 mesh

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$176.00

$176.00


Estimated to ship on29 January 2025Details


Request a Bulk Order

Select a Size

Change View
1 KG
$176.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
SB.52

$176.00


Estimated to ship on29 January 2025Details


Request a Bulk Order

Product Name

Amberlite IRA-96 free base, free base

description

weakly basic

form

beads

parameter

100 °C max. temp.

moisture

57-63%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (macroporous)

matrix active group

polyamine functional group

particle size

20-27 mesh
550-750 μm

operating pH

0-7

capacity

1.25 meq/mL by wetted bed volume

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

Application

Weakly basic anion exchange resin, useful for deionization, chromate recovery, formaldehyde deacidification, ammonium nitrate removal and recovery. Does NOT meet FDA Food Additive Regulation 21 CFR 173.25.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. 文献に「Amberlite IRA93」と表記されているのですが、「Amberlite™ IRA-96 遊離塩基」と異なるのでしょうか。異なるのであれば、どのような点が異なっているのでしょうか。

    1 answer
    1. There is no functional difference between Amberlite™ IRA93 and Amberlite™ IRA96. This product was initially listed as the IRA93 version and converted to IRA96 in 2000. While some parameters differ slightly these two versions are interchangeable. Note that the Amberlite™ IRA-96 has a particle size range of 0.55 - 0.75 mm while the Amberlite™ IRA93 particle size range was 0.4 - 0.5 mm. Amberlite™ IRA93 is no longer available.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service