Skip to Content
Merck
All Photos(1)

Key Documents

06433

Supelco

Amberlite IRA-410 Ion Exchange Resin

chloride form, 20-25 mesh, matrix styrene/divinylbenzene (gel)

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$280.00

$280.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
$280.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
SB.52

$280.00


Check Cart for Availability

Request a Bulk Order

Product Name

Amberlite IRA-410 chloride form, chloride form, matrix styrene/divinylbenzene (gel), 20-25 mesh

vapor density

<1 (vs air)

description

strongly basic

form

beads

autoignition temp.

800 °F

parameter

40 °C OH- form max. temp.
77 °C Cl- form max. temp.

moisture

45-51%

technique(s)

LPLC: suitable

loss

~45% loss on drying, 110 °C

matrix

styrene/divinylbenzene (gel)

matrix active group

benzyldimethyl(2-hydroxyethyl)ammonium functional group

particle size

20-25 mesh
600-750 μm

operating pH

0-14

capacity

1.25 meq/mL by wetted bed volume

separation technique

anion exchange

InChI

1S/C13H20NO.C10H10.ClH/c1-4-12-7-5-6-8-13(12)11-14(2,3)9-10-15;1-3-9-7-5-6-8-10(9)4-2;/h4-8,15H,1,9-11H2,2-3H3;3-8H,1-2H2;1H/q+1;;/p-1

InChI key

DPRMXNMXCFHCAS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Amberlite IRA-410 chloride form may be used as a column material for the purification of crude lipase obtained from Bacillus circulans[1] and extra-cellular lipase from Bacillus licheniformis MTCC 6824[2] by anion-exchange chromatography. It may also be used as an ion exchange resin for the recovery of amylases from maize malt by using expanded bed chromatography.[3]
Strongly basic (type II) anion exchange resin for water conditioning, removing iodine ions, neutralization of solutions, capturing xanthan and xanthene dyes, etc.; better regeneration efficiency than type I base resin.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Preparation of eicosapentaenoic acid concentrates from sardine oil by Bacillus circulans lipase.
Chakraborty K, et al.
Food Chemistry, 120(2), 433-442 (2010)
An extra-cellular alkaline metallolipase from Bacillus licheniformisMTCC 6824: purification and biochemical characterization.
Chakraborty Kajal and Raj RP
Food Chemistry, 109(4), 727-736 (2008)
Study of amylases recovery from maize malt by ion-exchange expanded bed chromatography.
Biazus JPM, et al.
Process Biochemistry (Oxford, United Kingdom), 41(8), 1786-1791 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service