Skip to Content
Merck
All Photos(1)

Key Documents

02890590

β-Asarone

primary reference standard

Synonym(s):

cis-1-Propenyl-2,4,5-trimethoxybenzene, cis-Asarone, cis-Isoasarone

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$643.00

$643.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$643.00

About This Item

Linear Formula:
(CH3O)3C6H2CH=CHCH3
CAS Number:
Molecular Weight:
208.25
Beilstein:
1910605
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$643.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

refractive index

n20/D 1.558 (lit.)

density

1.073 g/mL at 25 °C (lit.)

application(s)

food and beverages

storage temp.

−20°C

SMILES string

[H]\C(C)=C(/[H])c1cc(OC)c(OC)cc1OC

InChI

1S/C12H16O3/c1-5-6-9-7-11(14-3)12(15-4)8-10(9)13-2/h5-8H,1-4H3/b6-5-

InChI key

RKFAZBXYICVSKP-WAYWQWQTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference standard in the analysis of herbal medicinal products.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service