Skip to Content
Merck
All Photos(1)

Key Documents

00442

Supelco

4-Aminoazobenzene

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

4-Phenylazoaniline

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$244.00

$244.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$244.00

About This Item

Linear Formula:
C6H5N=NC6H4NH2
CAS Number:
Molecular Weight:
197.24
Colour Index Number:
11000
Beilstein:
1913042
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$244.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

bp

>360 °C (lit.)

mp

123-126 °C (lit.)

storage temp.

2-8°C

SMILES string

Nc1ccc(cc1)\N=N/c2ccccc2

InChI

1S/C12H11N3/c13-10-6-8-12(9-7-10)15-14-11-4-2-1-3-5-11/h1-9H,13H2/b15-14+

InChI key

QPQKUYVSJWQSDY-CCEZHUSRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
Aminoazobenzenes (AABs) are a class of compounds containing the azo group, which renders them colored. They have been used as dyes for fabrics, polishes, and food. 4-dimethylaminoazobenzene (DMAB), was used to color margarine.

Application

It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques.
The 4-Aminoazobenzene CRM can also be used as given below:

  • To study the spectral characteristics of 1-(2,4-diamino phenylazo) naphthalene and 4-aminoazobenzene by tautomerization and inclusion complexation
  • To study the inclusion complexes of ß-cyclodextrin (ß-CD) with 4-hydroxyazobenzene and 4-aminoazobenzene
  • In the detection of 4-aminoazobenzene using copper nanoclusters prepared using ascorbic acid

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Azonium-Ammonium Tautomerism and Inclusion Complexation of 1-(2, 4-diamino phenylazo) Naphthalene and 4-aminoazobenzene
Venkatesh G, et al.
Journal of Fluorescence, 21(4), 1485-1497 (2011)
Assembly behavior of inclusion complexes of ?-cyclodextrin with 4-hydroxyazobenzene and 4-aminoazobenzene
Liu Y, et al.
Organic & Biomolecular Chemistry, 584-591 null
Highly Luminescent Copper Nanoclusters Stabilized by Ascorbic Acid for the Quantitative Detection of 4-Aminoazobenzene
Li Q, et al.
Nanomaterials, 10(8), 1531-1531 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service