Skip to Content
Merck
All Photos(1)

Documents

30-2840

Sigma-Aldrich

Titanium(IV) butoxide

CP

Synonym(s):

Orthotitanic acid tetrabutylester, TNBT, TYZOR® TBT organic titanate, Tetrabutyl orthotitanate, Tetrabutyl titanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ti(OCH2CH2CH2CH3)4
CAS Number:
Molecular Weight:
340.32
Beilstein:
4148236
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:

grade

CP

form

liquid

reaction suitability

reagent type: catalyst
core: titanium

availability

available only in Japan

refractive index

n20/D 1.491 (lit.)

bp

206 °C/10 mmHg (lit.)

density

1.00 g/mL at 20 °C (lit.)

storage temp.

15-25°C

SMILES string

CCCCO[Ti](OCCCC)(OCCCC)OCCCC

InChI

1S/4C4H9O.Ti/c4*1-2-3-4-5;/h4*2-4H2,1H3;/q4*-1;+4

InChI key

YHWCPXVTRSHPNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used to prepare nanosized titania in the anatase form and ferroelectric Bi4Ti3O12 thin films. Used in the preparation of nanocrystalline TiO2 powders at room temperature.

Legal Information

TYZOR is a registered trademark of E. I. du Pont de Nemours and Company

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

136.8 °F

Flash Point(C)

58.2 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Anantharaj et al.
Biomacromolecules, 13(8), 2446-2455 (2012-06-21)
A new dual ester-urethane melt condensation methodology for biological monomers-amino acids was developed to synthesize new classes of thermoplastic polymers under eco-friendly and solvent-free polymerization approach. Naturally abundant L-amino acids were converted into dual functional ester-urethane monomers by tailor-made synthetic
Jun-Bing Fan et al.
Chemical communications (Cambridge, England), 46(31), 5805-5807 (2010-07-01)
A low cost and environmentally benign process for tetrabutyl titanate-controlled ring-opening polymerization of epsilon-caprolactone at ambient temperature of 10-40 degrees C is introduced affording poly(epsilon-caprolactone) with pre-designed molecular weights and narrow molecular weight distribution.
Huaqiang Zhuang et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 16(7), 1352-1355 (2015-02-26)
Ternary core-shell heterostructured rutile@anatase@Crx Oy nanorod arrays were elaborately designed as photoanodes for efficient photoelectrochemical water splitting under visible-light illumination. The four-fold enhanced and stabilized visible-light photocurrent highlights the unique role of the interim anatase layer in accelerating the interfacial
Jun Lu et al.
Nature communications, 5, 5693-5693 (2014-12-17)
The capacity fade of lithium manganate-based cells is associated with the dissolution of Mn from cathode/electrolyte interface due to the disproportionation reaction of Mn(III), and the subsequent deposition of Mn(II) on the anode. Suppressing the dissolution of Mn from the
Mukesh K Sharma et al.
Biosensors & bioelectronics, 74, 299-304 (2015-07-08)
Bacillus anthracis is a bioterrorism agent classified by the Centers for Disease Control and Prevention (CDC). Herein, a novel electrochemical immunosensor for the sensitive, specific and easy detection of anthrax protective antigen (PA) toxin in picogram concentration was developed. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service