Skip to Content
Merck
All Photos(1)

Key Documents

28-1400

Sigma-Aldrich

Silver(I) oxide

SAJ first grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ag2O
CAS Number:
Molecular Weight:
231.74
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
grade:
SAJ first grade
form:
solid

grade

SAJ first grade

Assay

≥98.0%

form

solid

availability

available only in Japan

storage temp.

15-25°C

SMILES string

[Ag]O[Ag]

InChI

1S/2Ag.O

InChI key

KHJDQHIZCZTCAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weijia Zhou et al.
Physical chemistry chemical physics : PCCP, 12(45), 15119-15123 (2010-10-22)
Electrostatic self-assembled Ag(2)O/TiO(2) nanobelts heterostructure was synthesized via simple physical mixing of Ag(2)O nanoparticles and TiO(2) nanobelts. The morphologies and microstructures of Ag(2)O/TiO(2) nanobelt heterostructure were characterized by high resolution transmission electron microscopy. The interface dominated high UV photocatalytic activity
Chiara Uboldi et al.
Mutation research, 745(1-2), 11-20 (2011-11-19)
Although amorphous silica nanoparticles (aSiO(2)NPs) are believed to be non-toxic and are currently used in several industrial and biomedical applications including cosmetics, food additives and drug delivery systems, there is still no conclusive information on their cytotoxic, genotoxic and carcinogenic
Silver stools revisited.
Jonathan Wyse et al.
Gastroenterology, 144(5), e9-e10 (2013-03-26)
Arixin Bo et al.
Journal of hazardous materials, 246-247, 199-205 (2013-01-15)
Emergency treatment of radioactive material leakage and safety disposal of nuclear waste is a constant concern all along with the development of radioactive materials applications. To provide a solution, titanate with large surface area (143 m(2)g(-1)) and a lamina morphology
Frédéric Louërat et al.
Organic & biomolecular chemistry, 9(6), 1768-1773 (2011-01-26)
The concomitant use of silver oxide and catalytic amount of TBAF allowed the efficient and chemoselective coupling of readily available 4-chloro- and 4-methyl-2-trimethyl-silyl-pyridines with heteroaromatic and aromatic halides. Based on control experiments, a mechanism involving the formation of a pyridylsilver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service