Skip to Content
Merck
All Photos(1)

Key Documents

223638

Sigma-Aldrich

Silver oxide

predominantly silver(II) oxide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$181.00
50 G
$622.00

$181.00


Estimated to ship on31 March 2025


Request a Bulk Order

Select a Size

Change View
10 G
$181.00
50 G
$622.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

$181.00


Estimated to ship on31 March 2025


Request a Bulk Order

description

mixture

Quality Level

form

powder and chunks

impurities

silver(I) oxide (remainder)

density

7.483 g/mL at 25 °C

SMILES string

O=[Ag]

InChI

1S/Ag.O

InChI key

OTCVAHKKMMUFAY-UHFFFAOYSA-N

Application

Converts benzyl halides directly into benzyl ethers.[1] Also used as a surface catalyst in the epoxidation of alkenes.[2]

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies in Surface Science and Catalysis, 59, 377-377 (1991)
Synthetic Communications, 23, 749-749 (1993)
Chiara Uboldi et al.
Mutation research, 745(1-2), 11-20 (2011-11-19)
Although amorphous silica nanoparticles (aSiO(2)NPs) are believed to be non-toxic and are currently used in several industrial and biomedical applications including cosmetics, food additives and drug delivery systems, there is still no conclusive information on their cytotoxic, genotoxic and carcinogenic
Woonsup Shin et al.
Analytical chemistry, 83(12), 5023-5025 (2011-05-10)
When a current or a voltage is applied across the ceramic membrane of the nongassing Ag/Ag(2)O-SiO(2)-Ag/Ag(2)O pump, protons produced in the anodic reaction 2Ag(s) + H(2)O → Ag(2)O(s) + 2H(+) + 2e(-) are driven to the cathode, where they are
Frédéric Louërat et al.
Organic & biomolecular chemistry, 9(6), 1768-1773 (2011-01-26)
The concomitant use of silver oxide and catalytic amount of TBAF allowed the efficient and chemoselective coupling of readily available 4-chloro- and 4-methyl-2-trimethyl-silyl-pyridines with heteroaromatic and aromatic halides. Based on control experiments, a mechanism involving the formation of a pyridylsilver

Articles

Oxidation and reduction reactions are some of the most common transformations encountered in organic synthesis

Questions

  1. Studiul comportamentului Ag2O la temperaturi cuprinse intre 150 și 170 de grade Celsius.

    1 answer
    1. Silver oxide has a melting point of 300°C. The nature or characteristics of this chemical between 150 and 170°C would need to be determined by the end user. It may be helpful to view the peer-reviewed articles associated with this product:
      https://www.sigmaaldrich.com/search/223638?focus=papers&page=1&perpage=30&sort=relevance&term=223638&type=citation_search

      To discuss further, please navigate to the link https://www.sigmaaldrich.com/techservice, and click on "Product Technical Inquiries" under the Products Section with all the required information so that a member of the Technical Service team can reach out to assist further. Thank you.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service