Skip to Content
Merck
All Photos(1)

Documents

28-0440

Sigma-Aldrich

Sebacoyl chloride

CP

Synonym(s):

Sebacyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO(CH2)8COCl
CAS Number:
Molecular Weight:
239.14
Beilstein:
1365665
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

CP

form

liquid

availability

available only in Japan

refractive index

n20/D 1.468 (lit.)

bp

168 °C/12 mmHg (lit.)

mp

−5-−3 °C (lit.)

density

1.121 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)CCCCCCCCC(Cl)=O

InChI

1S/C10H16Cl2O2/c11-9(13)7-5-3-1-2-4-6-8-10(12)14/h1-8H2

InChI key

WMPOZLHMGVKUEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Poncelet et al.
Journal of microencapsulation, 11(1), 31-40 (1994-01-01)
A microencapsulation technique is proposed involving the formation of a polyethyleneimine (PEI) membrane crosslinked by an acid dichloride. The membranes were formed at pH 8 in a non-polar solvent, conditions which are better suited for the encapsulation of biocatalysts or
Marianna Sikorska et al.
BioFactors (Oxford, England), 18(1-4), 173-183 (2003-12-30)
We have derivatised alpha-tocopherol (vitamin E) to a water-soluble polyoxyethanyl-alpha - tocopheryl sebacate (PTS) and discovered that it formed a non-covalent complex with CoQ10 at a molar ratio of 2:1 (PTS-CoQ10). This complex was water-soluble and remained stable for extended
Zhenhua Xu et al.
International journal of pharmaceutics, 426(1-2), 182-192 (2012-01-24)
In this study we describe a novel polymer, mPPS-FA, synthesized as a potential gene transfer vector. To complete mPPS-FA, folic acid was conjugated to a backbone (named mPPS) consisting of a copolymer of methyl PEG-2000, PEI-600, and sebacoyl chloride. (1)H
P L Madan et al.
Pharmaceutical research, 6(8), 714-718 (1989-08-01)
The effect of selected variables on the release characteristics of theophylline microcapsules prepared with 1,6-hexamethylene diamine and sebacoyl chloride via interfacial polycondensation has been investigated. The nature of the microcapsules and the release characteristics of theophylline from the microcapsules were
I Montasser et al.
International journal of pharmaceutics, 335(1-2), 176-179 (2006-12-19)
Formulation of nanocapsules based on polyureas and polyamides have been tested using a patented process. This method based on polycondensation reaction of two complementary monomers and spontaneous formation of oil in water emulsion, is an alternative concept to the known

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service