Skip to Content
Merck
All Photos(1)

Key Documents

52003

Supelco

Hexamethylenediamine

analytical standard

Synonym(s):

1,6-Diaminohexane, 1,6-Hexanediamine, HMDA

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$135.00

$135.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$135.00

About This Item

Linear Formula:
NH2(CH2)6NH2
CAS Number:
Molecular Weight:
116.20
Beilstein:
1098307
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21

$135.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

4 (vs air)

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

expl. lim.

6.3 %

refractive index

n20/D 1.439 (lit.)

mp

40-45 °C
42-45 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

SMILES string

NCCCCCCN

InChI

1S/C6H16N2/c7-5-3-1-2-4-6-8/h1-8H2

InChI key

NAQMVNRVTILPCV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service