Skip to Content
Merck
All Photos(1)

Key Documents

15-1450

Sigma-Aldrich

Iron standard solution

suitable for atomic absorption spectrometry, 1 mg/mL Fe, 1000 ppm Fe

Synonym(s):

Iron(III) nitrate solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

form

liquid

availability

available only in Japan

concentration

1 mg/mL Fe
1000 ppm Fe

suitability

suitable for atomic absorption spectrometry

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Ndong et al.
International endodontic journal, 45(6), 557-564 (2012-01-25)
To determine the effect of increasing the proportion of zinc on partially stabilized cement (PSC) produced using a one-step sol gel process. A one-step sol-gel process of Portland cement-based PSC with Zn was synthesized by replacing iron nitrate. The crystalline
Marek Basler et al.
Proteomics, 6(23), 6194-6206 (2006-11-30)
Restricting bacterial growth by iron-chelating proteins that reduce iron availability in mucosal secretions and body fluids belongs to basic mechanisms of innate immunity. Most pathogens and commensals thus developed gene regulons responding to iron concentration and encoding iron acquisition systems
Runjuan Wang et al.
Bioresource technology, 120, 290-294 (2012-07-10)
The study investigated the effects of two-step pretreatment on fermentable sugar production from corn stalk silage. In the first step, the corn stalk silage was extracted by tepid water and then the solid was pretreated using Fe(NO(3))(3) as catalyst. The
Mara Ghiazza et al.
Chemical research in toxicology, 24(1), 99-110 (2010-12-22)
The mechanism of enhancement/inhibition of quartz toxicity induced by iron is still unclear. Here the amount of iron on a fibrogenic quartz (Qz) was increased by wet impregnation (Fe(NO(3))(3) 0.67 and 6.7 wt %). X-ray diffraction (XRD), XRF diffuse reflectance
Youshan Sun et al.
Applied biochemistry and biotechnology, 164(6), 918-928 (2011-02-15)
Corn stover silage is an attractive raw material for the production of biofuels and chemicals due to its high content of carbohydrates and easy degradability. The effects of Fe(NO(3))(3) pretreatment conditions on sugar yields were investigated for corn stover silage.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service