Skip to Content
Merck
All Photos(1)

Key Documents

15-0685

Sigma-Aldrich

Iodine trichloride

SAJ special grade, ≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ICl3
CAS Number:
Molecular Weight:
233.26
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
SAJ special grade
form:
solid
Pricing and availability is not currently available.

grade

SAJ special grade

Assay

≥97.0%

form

solid

availability

available only in Japan

SMILES string

Cl[I](Cl)Cl

InChI

1S/Cl3I/c1-4(2)3

InChI key

PAWIVBWALDNUJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service