Skip to Content
Merck
All Photos(1)

Key Documents

12-0240

Sigma-Aldrich

Gelatin from bovine skin

SAJ first grade

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
Pricing and availability is not currently available.

grade

SAJ first grade

availability

available only in Japan

Looking for similar products? Visit Product Comparison Guide

Components

Gelatin is a heterogeneous mixture of water-soluble proteins of high average molecular masses, present in collagen. Proteins are extracted by boiling the relevant skin, tendons, ligaments, bones, etc. in water. Type A gelatin is derived from acid-cured tissue. Type B is derived from lime-cured tissue.

Caution

Dry gelatin, when stored in airtight containers at room temperature, will remain unchanged for many years. When heated at 100°C in the presence of air, it swells becomes soft and disintegrates to a carbonaceous mass with evolution of pyridine bases and ammonia.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frank Galka et al.
Infection and immunity, 76(5), 1825-1836 (2008-02-06)
Secretion of effector molecules is one of the major mechanisms by which the intracellular human pathogen Legionella pneumophila interacts with host cells during infection. Specific secretion machineries which are responsible for the subfraction of secreted proteins (soluble supernatant proteins [SSPs])
L Du et al.
Poultry science, 92(9), 2463-2474 (2013-08-21)
Gelatins were prepared from chicken and turkey heads in a series of batch extractions at 2 different temperatures (50 and 60°C), and their composition and functional properties were evaluated. Gelatin yield from chicken and turkey heads was 52.29 and 62.76%
Melika Sarem et al.
Carbohydrate polymers, 93(2), 635-643 (2013-03-19)
In the engineering of soft tissues, designing the scaffolds with adequate strength and controllable biodegradation properties are essential. To fulfill such design criteria gelatin/carbohydrate chitosan (G/CC) scaffolds have gained much attention in engineering replacement of soft tissues. In this research
Pui Khoon Hong et al.
Food chemistry, 142, 285-293 (2013-09-05)
A mixture of novel glycopeptides from glycosylation between cold water fish skin gelatin hydrolysates and glucosamine (GlcN) via transglutaminase (TGase), as well as glycation between fish gelatin hydrolysate and GlcN were identified by their pattern of molecular distribution using MALDI-TOF-MS.
Somayeh Hallaj Nezhadi et al.
Journal of drug targeting, 17(10), 731-738 (2009-10-30)
Gelatin is a natural, biocompatible, nontoxic, edible, and inexpensive macromolecule. These properties result in its wide application in pharmaceutical, medical, and cosmetic products. Recently, it has been used for the delivery of such gene therapeutic entities as plasmid DNA. This

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service