Skip to Content
Merck
All Photos(1)

Key Documents

L9655

Sigma-Aldrich

Linoleic Acid-Oleic Acid-Albumin, 100x

liquid, sterile-filtered, BioReagent, suitable for cell culture

Synonym(s):

Fatty Acid-Albumin-complexes

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$75.40

$75.40


Available to ship on09 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 ML
$75.40

About This Item

UNSPSC Code:
12352211
NACRES:
NA.75

$75.40


Available to ship on09 April 2025Details


Request a Bulk Order

biological source

protein from bovine

sterility

sterile-filtered

product line

BioReagent

form

liquid

composition

linoleic acid and oleic acid, 2 mol/mol albumin (each)

concentration

100 mg/mL BSA in DPBS

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

storage temp.

2-8°C

General description

Fatty Acid-Albumin-complexes have been employed as alternative sources of lipids in the development of serum-free media. Fatty acids bind to serum proteins in high proportions. Such proteins may release beneficial fatty acids and bind those that are inhibitory. Oleic acid bound to bovine serum albumin (BSA) is beneficial to the growth of a variety of cell types (e.g., baby hamster kidney (BHK) hybridoma). A mixture of poly-unsaturated and monosaturated fatty acids (i.e., linoleic acid and oleic acid) produces synergistic effect when used as a media supplement.

Application

Linoleic Acid-Oleic Acid-Albumin, 100x has been used:
  • to induce acute hepatocellular stress in primary hepatocytes [1]
  • to induce oxidative stress in cytotrophoblasts[2]
  • to induce lipid droplet accumulation in HepG2 cells[3]

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Moody et al.
Biochimica et biophysica acta. Gene regulatory mechanisms, 1862(2), 141-152 (2019-01-04)
Carnitine palmitoyltransferase 1 (Cpt1a) is a rate-limiting enzyme that mediates the transport of fatty acids into the mitochondria for subsequent beta-oxidation. The objective of this study was to uncover how diet mediates the transcriptional regulation of Cpt1a. Pregnant Sprague Dawley
Pierluigi Ramadori et al.
Free radical biology & medicine, 91, 114-126 (2015-12-25)
Generation of reactive oxygen species (ROS) in response to fatty acids accumulation has been classically proposed as a possible "second hit" triggering progression from simple steatosis to non-alcoholic steatohepatitis (NASH). In this study we challenged hepatocyte-specific Keap1 knockout mice (Keap1(Δhepa))
Anna Virginia Adriana Pirozzi et al.
Antioxidants (Basel, Switzerland), 10(1) (2021-01-06)
Several plant extracts are acquiring increasing value because of their antioxidant activity and hypolipidemic properties. Among them, great interest has been recently paid to açai fruit as a functional food. The aim of this study was to test the ability
Yukimasa Takeda et al.
Scientific reports, 10(1), 3775-3775 (2020-03-01)
Brown adipocytes coordinate systemic energy metabolism associated with the pathogenesis of obesity and related metabolic diseases including type 2 diabetes. We have previously reported chemical compound-induced brown adipocytes (ciBAs) converted from human dermal fibroblasts without using transgenes. In this study
Liang Tian et al.
International journal of molecular sciences, 20(5) (2019-03-13)
This study aimed to characterize the full-length cDNA of MARK4 in Sus scrofa, and evaluated its potential role in the regulation of lipid accumulation in pig placental trophoblasts and analyzed signaling pathways involved, thereby providing insights into mechanisms for placental

Articles

Importance and uses of linoleic acid in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

How the unsaturated fatty acid, oleic acid and other cell culture components affect the performance of serum-free, protein-free cell culture systems used for biomanufacturing heterologous proteins including monoclonal antibodies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service