Skip to Content
Merck
All Photos(1)

Key Documents

05-6914

Sigma-Aldrich

Cyanide standard solution

1 mg/mL CN-

Synonym(s):

Potassium cyanide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CKN
CAS Number:
Molecular Weight:
65.12
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

form

liquid

availability

available only in Japan

concentration

1 mg/mL CN-

storage temp.

2-8°C

SMILES string

[K]C#N

InChI

1S/CN.K/c1-2;

InChI key

YUZRZFQHUCKACF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Zhang Yu et al.
Journal of hazardous materials, 225-226, 190-194 (2012-05-29)
A study was conducted to investigate activities of nitrate reductase (NR) and glutamine synthetase (GS) in plants during cyanide metabolism. Young rice seedlings (Oryza sativa L. cv. XZX 45) were grown in the nutrient solutions containing KNO(3) or NH(4)Cl and
Hajar Sahebalzamani et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 90, 218-222 (2012-02-22)
Two new aluminate compounds was prepared by the reaction of AlCl(3) with KX (X=SCN(-), CN(-)) in a 1:1 mole ratio. In these salts the aluminum atom is surrounded by three chlorine atoms and a ligand (X=SCN(-), CN(-)). In AlCl(3)SCN anion
Fei Xu et al.
Plant, cell & environment, 35(11), 1983-1997 (2012-05-05)
The effects of potassium cyanide (KCN) pretreatment on the response of cucumber (Cucumis sativus L.) plants to salt, polyethylene glycol (PEG) and cold stress were investigated in the present study. Here, we found that KCN pretreatment improved cucumber seedlings tolerance
M V Dolgushin et al.
Meditsina truda i promyshlennaia ekologiia, (3)(3), 36-40 (2013-06-22)
Cytochemical analysis of acid phosphatase was used to evaluate lysosomal membranes stability under oral intake of potassium cyanide by rats over one month in daily doses of 1.30 mg/kg (1/10 LD50) and 0.65 mg/kg (1/20 LD50). The authors demonstrated phase-related
Paisarn Khawsak et al.
Parasitology research, 110(2), 629-636 (2011-07-29)
The Cu/Zn superoxide dismutase gene from Wuchereria bancrofti (Cu/Zn WbSOD) was isolated by PCR using degeneracy primers. The complete Cu/Zn WbSOD consisted of 1,032 nucleotides containing 4 exons (477 nucleotides) and 3 introns. The molecular phylogenetic analysis of the Cu/Zn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service