Merck
  • Density functional theory studies of structural properties, energies and natural band orbital for two new aluminate compounds.
Density functional theory studies of structural properties, energies and natural band orbital for two new aluminate compounds.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-02-22)
Hajar Sahebalzamani, Shahriare Ghammamy, Kheyrollah Mehrani, Shahram Jahandide, Farshid Salimi
ABSTRACT
Two new aluminate compounds was prepared by the reaction of AlCl(3) with KX (X=SCN(-), CN(-)) in a 1:1 mole ratio. In these salts the aluminum atom is surrounded by three chlorine atoms and a ligand (X=SCN(-), CN(-)). In AlCl(3)SCN anion, the SCN coordinates to the Al through sulfur and AlCl(3)CN anion the CN(-) coordinates to the Al center through carbon. The molecular geometry, vibrational frequencies, energies and natural bond orbital (NBO) in the ground state are calculated by using the DFT (B3LYP) methods with 6-311G* basis sets. The geometries and normal modes of vibrations obtained from B3LYP calculations are in good agreement with the experimentally observed data.
Sigma-Aldrich
Potassium cyanide-15N, 98 atom % 15N
Sigma-Aldrich
Potassium cyanide, BioUltra, ≥98.0% (AT)
Sigma-Aldrich
Potassium cyanide, ACS reagent, ≥96.0%
Sigma-Aldrich
Potassium cyanide, technical, ≥96%