Skip to Content
Merck
All Photos(1)

Key Documents

03-3790

Sigma-Aldrich

Bromocresol Purple

SAJ special grade

Synonym(s):

5,5′-Dibromo-o-cresolsulfonphthalein, Bromcresol purple sultone form

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H16Br2O5S
CAS Number:
Molecular Weight:
540.22
Beilstein:
359618
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

SAJ special grade

availability

available only in Japan

pH

5.2-6.8, yellow to purple

mp

240 °C (dec.) (lit.)

SMILES string

Cc1cc(cc(Br)c1O)C2(OS(=O)(=O)c3ccccc23)c4cc(C)c(O)c(Br)c4

InChI

1S/C21H16Br2O5S/c1-11-7-13(9-16(22)19(11)24)21(14-8-12(2)20(25)17(23)10-14)15-5-3-4-6-18(15)29(26,27)28-21/h3-10,24-25H,1-2H3

InChI key

ABIUHPWEYMSGSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Spectrophotometric determination of serum-albumin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Faizah Mohd Faizul et al.
Journal of photochemistry and photobiology. B, Biology, 90(1), 1-7 (2007-11-21)
The interaction between bromocresol purple (BCP) and bovine serum albumin (BSA)/porcine serum albumin (PSA) was investigated both in the absence and presence of bilirubin (BR) using absorption/absorption difference spectroscopy. A significant red shift in the absorption maxima of BCP accompanied
Munzer Kattan et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 69(2), 377-380 (2010-11-27)
Polyvinyl chloride (PVC) dyed with bromocresol purple was investigated as a high-dose radiation dosimeter. The absorbance at 417 nm depends linearly on the dose below 50 kGy. The response depends neither on dose rate nor on the irradiation temperature. The
M Arab Chamjangali et al.
Journal of hazardous materials, 166(2-3), 701-705 (2009-01-13)
A simple, selective, and sensitive kinetic spectrophotometric method is described for the determination of trace amounts of phenylhydrazine, which is based on its inhibition effect on the reaction between meta cresol purple (MCP) and periodate in the presence of bromide
C H White et al.
Journal of dairy science, 89(7), 2459-2464 (2006-06-15)
The MicroFoss method was evaluated for its effectiveness as an indicator of fluid milk shelf life. Half-gallon, 2% fat fluid milk samples (n = 90) were obtained from a milk processing plant on 3 occasions postpasteurization and evaluated for shelf
André Klauss et al.
Photosynthesis research, 102(2-3), 499-509 (2009-03-31)
Determination of thermodynamic parameters of water oxidation at the photosystem II (PSII) manganese complex is a major challenge. Photothermal beam deflection (PBD) spectroscopy determines enthalpy changes (ΔH) and apparent volume changes which are coupled with electron transfer in the S-state

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service