Skip to Content
Merck
All Photos(1)

Documents

01-6010

Sigma-Aldrich

Potassium antimonyl tartrate trihydrate

JIS special grade, ≥99.5%

Synonym(s):

Antimony potassium tartrate trihydrate, Tartar emetic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4K2O12Sb2 · 3H2O
CAS Number:
Molecular Weight:
667.87
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.5%

form

powder

availability

available only in Japan

pH

4 (20 °C)

mp

≥300 °C (lit.)

SMILES string

O.O.O.[K+].[K+].O=C1O[Sb-]23OC1C4O[Sb-]5(OC(C(O2)C(=O)O3)C(=O)O5)OC4=O

InChI

1S/2C4H4O6.2K.3H2O.2Sb/c2*5-1(3(7)8)2(6)4(9)10;;;;;;;/h2*1-2H,(H,7,8)(H,9,10);;;3*1H2;;/q2*-2;2*+1;;;;2*+3/p-4

InChI key

WBTCZEPSIIFINA-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zahra Faghiri et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 23(8), 2780-2789 (2009-04-15)
Schistosomes are parasitic platyhelminths that constitute an important public health problem globally. Infection is characterized by the presence of adult worms within the vasculature of their hosts, where they can reside for many years. The worms are covered by an
Daniel B Liarte et al.
Parasitology research, 107(1), 205-212 (2010-04-08)
In the present study, we selected in vitro populations of Leishmania Viannia guyanensis, L.V. braziliensis, L. Leishmania amazonensis and L.L. infantum chagasi that were resistant to potassium antimony tartrate (SbIII). The resistance index of these populations varied from 4- to
Mostafa K Mohamed et al.
Journal of medical virology, 78(9), 1185-1189 (2006-07-19)
The origin of the hepatitis C virus (HCV) epidemic in Egypt has been attributed to intravenous schistosomiasis treatment in rural areas in the 1960s to 70s. The objective of this study was to estimate the HCV-related morbidity in a rural
Christian Brochu et al.
Cell stress & chaperones, 9(3), 294-303 (2004-11-17)
Antimony-containing drugs are still the drugs of choice in the treatment of infections caused by the parasite Leishmania. Resistance to antimony is now common in some parts of the world, and several mechanisms of resistance have been described. By transfecting
Avijit Sarkar et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 79(1), 35-45 (2010-12-25)
Nitric oxide (NO) has been demonstrated to be a principal effector molecule responsible for mediating intracellular killing of Leishmania parasites, the causative organism of leishmaniasis. As measurement of intracellular NO remains a challenge to biologists, we have developed a flow

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service