Merck
HCV-related morbidity in a rural community of Egypt.
Journal of medical virology (2006-07-19)
Mostafa K Mohamed, Iman Bakr, Mostafa El-Hoseiny, Naglaa Arafa, Abubakr Hassan, Soheir Ismail, Mohamed Anwar, Mohamed Attala, Claire Rekacewicz, Khaled Zalata, Mohamed Abdel-Hamid, Gamal Esmat, Arnaud Fontanet
ABSTRACT
The origin of the hepatitis C virus (HCV) epidemic in Egypt has been attributed to intravenous schistosomiasis treatment in rural areas in the 1960s to 70s. The objective of this study was to estimate the HCV-related morbidity in a rural area where mass schistosomiasis treatment campaigns took place 20-40 years before. The study sample included 2,425 village residents aged 18-65 years recruited through home-based visits. Overall, HCV antibody prevalence was 448/2,425 = 18.5% (95% CI = 16.9-20.1%), reaching 45% in males over 40 years, and 30% in females over 50 years. Of those with HCV antibodies, 284/448 (63.4%, 95% CI = 58.7-67.9%) had chronic HCV infection, among which 107/266 (40.2%, 95% CI = 34.3-46.4%) had elevated alanine aminotransferase (ALT). As part of pre-treatment screening, 26 consenting patients had a liver biopsy: 13 (50.0%) had a treatment indication. Thus, of all patients with HCV antibodies, 13 (2.9%) were eligible for treatment and willing to be treated. The relatively low level of morbidity observed in this study is discussed in view of co-factors of HCV infection progression, such as young age at infection, absence of alcohol intake, the prevalence of Schistosoma mansoni infection, and the prevalence of chronic hepatitis B.
Sigma-Aldrich
Potassium antimonyl tartrate trihydrate, purum p.a., 99.0-103% (RT)
Sigma-Aldrich
Potassium antimonyl tartrate trihydrate, puriss., meets analytical specification of USP, 99.0-103.0%, powder
Sigma-Aldrich
Potassium antimonyl tartrate trihydrate, ACS reagent, ≥99%