Skip to Content
Merck
All Photos(1)

Documents

01-5200

Sigma-Aldrich

Ammonium thiocyanate

SAJ first grade, ≥99%

Synonym(s):

Ammonium rhodanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4SCN
CAS Number:
Molecular Weight:
76.12
Beilstein:
3595135
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:

grade

SAJ first grade

vapor pressure

0.000114 hPa ( 20 °C)

Assay

≥99%

form

solid

availability

available only in Japan

pH

4.0-5.5 (25 °C, 76.1 g/L)

mp

152-154 °C (lit.)

SMILES string

N.SC#N

InChI

1S/CHNS.H3N/c2-1-3;/h3H;1H3

InChI key

SOIFLUNRINLCBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fumi Miyagawa et al.
Journal of immunology (Baltimore, Md. : 1950), 181(2), 1109-1119 (2008-07-09)
To elucidate the mechanisms controlling peripheral tolerance, we established two transgenic (Tg) mouse strains expressing different levels of membrane-bound OVA (mOVA) as a skin-associated self-Ag. When we transferred autoreactive TCR-Tg CD8 T cells (OT-I cells), keratin 14 (K14)-mOVA(high) Tg mice
Xiaolian Gu et al.
The Journal of investigative dermatology, 135(8), 2077-2083 (2015-04-02)
Epigenetic modifications by DNA methylation are associated with a wide range of diseases. Previous studies in psoriasis have concentrated on epigenetic changes in immune cells or in total skin biopsies that include stromal-associated changes. In order to improve our understanding
Kamlesh Shrivas et al.
Journal of hazardous materials, 161(1), 325-329 (2008-05-09)
A novel and sensitive spectrophotometric method for the determination of molybdenum at trace levels in environmental and biological samples is proposed. The method is based on the reaction of Mo (V) with thiocyanate (SCN(-)) and methyltrioctyl ammonium chloride (MTOAC) in
Anders Clemmensen et al.
Experimental dermatology, 18(11), 979-984 (2009-08-04)
To obtain a separation of the epidermal and dermal compartments to examine compartment specific biological mechanisms in the skin, we incubated 4 mm human skin punch biopsies in ammonium thiocyanate. We wanted to test (i) the histological quality of the
G Almanzar et al.
Journal of immunological methods, 387(1-2), 36-42 (2012-10-02)
Antibody avidity, defined as the strength of binding between antibody and antigen, represents a functional measure of affinity maturation of antibodies. Determination of the antibody avidity is usually performed separating high and low avidity antibodies by dissociating agents, but measurement

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service