Skip to Content
Merck
All Photos(1)

Key Documents

SAE0192

Sigma-Aldrich

Parathyroid Hormone (1-84) Human

Recombinant expressed in E. coli

Synonym(s):

PTH, Parathormone

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$557.00

$557.00


Check Cart for Availability


Select a Size

Change View
100 μG
$557.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77
biological source:
Escherichia coli
form:
solid

$557.00


Check Cart for Availability

biological source

Escherichia coli

Quality Level

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

solid

shipped in

ambient

storage temp.

−20°C

General description

Parathyroid hormone (PTH), or parathormone, is secreted by the parathyroid glands as a polypeptide containing 84 amino acids. It acts to increase the concentration of calcium in the blood, whereas calcitonin (a hormone produced by the parafollicular cells of the thyroid gland) acts to decrease calcium concentration.
Parathyroid Hormone 1-84 (full length) Human Recombinant produced in E. coli is a single, non-glycosylated, polypeptide chain containing 84 amino acids (MW of ∼9.4kDa) with the following amino acid sequence:
SVSEIQLMHN LGKHLNSMER VEWLRKKLQD VHNFVALGAP LAPRDAGSQR PRKKEDNVLV ESHEKSLGEA DKADVNVLTK AKSQ

Biochem/physiol Actions

PTH acts to increase the concentration of calcium in the blood by acting upon parathyroid hormone receptor in three parts of the body: In the bones- It enhances the release of calcium from the large reservoir contained in the bones. Bone resorption is the normal destruction of bone by osteoclasts, which are indirectly stimulated by PTH. Recombinant Human full length PTH 1-84 has potential as an anti-osteoporotic agent, due to its properties as a bone formation stimulant, it increases bone turnover, stimulating osteoblasts and reducing both vertebral and non-vertebral fractures.

Physical form

The product is supplied as lyophilized powder, from a 0.2μm filtered concentrated solution in PBS pH7.4.

Storage and Stability

It is recommended to reconstitute the lyophilized PTH in sterile purified water, at not less than 100 μg/ml, which can then be further diluted to other aqueous solutions.
Upon reconstitution PTH should be stored at 2-8 °C up to 7 days and for future use at -20°C. For long term storage it is recommended to add a carrier protein (0.1% HSA or BSA). Avoid freeze-thaw cycles

Disclaimer

This product is for R&D use only, not for drug, household, or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service