EHIFIDNTP-RO
Roche
Expand™ High Fidelity PCR System, dNTPack
suitable for PCR, High Fidelity PCR, hotstart: no, dNTPs included
Select a Size
$252.00
Select a Size
About This Item
$252.00
Recommended Products
usage
sufficient for ≤1,000 reactions (04738276001)
sufficient for ≤200 reactions (04738268001)
sufficient for ≤40 reactions (04738250001)
Quality Level
feature
High Fidelity PCR
dNTPs included
hotstart: no
packaging
pkg of 2,500 U (04738276001 [10x250 U])
pkg of 100 U (04738250001)
pkg of 500 U (04738268001 [2x250 U])
manufacturer/tradename
Roche
technique(s)
PCR: suitable
input
purified DNA
storage temp.
−20°C
Related Categories
General description
Application
- PCR
- RT-PCR
Features and Benefits
- Use one enzyme for all applications. Enjoy high yield, fidelity, and flexibility.
- Detect PCR products previously undetectable and avoid false negatives.
- Achieve successful results from small quantities of template DNA.
Packaging
Quality
Unit Definition
Each dNTPack contains 10mM additive-free sodium salt nucleotides in a ready-to-use mix.
Other Notes
Legal Information
Kit Components Only
- Expand High Fidelity Enzyme mix
- Expand High Fidelity Buffer, with 15 mM MgCl2 10x concentrated
- Expand High Fidelity Buffer, without MgCl2
- MgCl2 Stock Solution 25 mM
- PCR Nucleotide Mix
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service