Skip to Content
Merck
All Photos(1)

Key Documents

11685929001

Sigma-Aldrich

Guanidine thiocyanate

Synonym(s):

Guanidine thiocyanate, Guanidinium rhodanide, Guanidinium thiocyanate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$493.00

$493.00


Check Cart for Availability


Select a Size

Change View
500 G
$493.00

About This Item

Linear Formula:
NH2C(=NH)NH2 · HSCN
CAS Number:
Molecular Weight:
118.16
Beilstein:
3563461
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

$493.00


Check Cart for Availability

form

solid

mol wt

118.16

packaging

pkg of 500 g

manufacturer/tradename

Roche

mp

120-122 °C (lit.)

absorption

≤0.10 at 300 and 410 nm in water at 6 M

shipped in

ambient

storage temp.

20-25°C

SMILES string

SC#N.NC(N)=N

InChI

1S/CH5N3.CHNS/c2-1(3)4;2-1-3/h(H5,2,3,4);3H

InChI key

ZJYYHGLJYGJLLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Guanidine thiocyanate is chaotropic salts which lyse cells and along with nuclease binding matrix it is also seen to inhibit nuclease activity during extraction of DNA. It works as a strong RNase inhibitor in extraction buffers.

Application

Chaotropic agent and strong denaturant; solubilizes cells.
Suitable as denaturing agent for proteins. Guanidine thiocyanate is a stronger denaturant than guanidine hydrochloride.
Suitable as denaturing agent for proteins. Guanidine thiocyanate is a stronger denaturant than guanidine hydrochloride.[1]

Specifications

Absorbance: A300 and A410 ≤0.10 (6 M in H2O) each
Formula: CH5N3 x HSCN

Physical form

Crystals

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 Inhalation - Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weidong Wu et al.
The Journal of biological chemistry, 278(30), 28258-28263 (2003-05-14)
The tumor suppressor PTEN is a putative negative regulator of the phosphatidylinositol 3-kinase/Akt pathway. Exposure to Zn2+ ions induces Akt activation, suggesting that PTEN may be modulated in this process. Therefore, the effects of Zn2+ on PTEN were studied in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service