Skip to Content
Merck
All Photos(1)

Key Documents

10662046001

Roche

β-Galactose Dehydrogenase S

suspension, ~80 units/mg protein (At 25 °C with galactose as the substrate.), pkg of 50 U

Synonym(s):

D-galactose:NAD+ 1-oxidoreductase

Sign Into View Organizational & Contract Pricing

Select a Size

50 UNITS
$570.00

$570.00


Check Cart for Availability


Select a Size

Change View
50 UNITS
$570.00

About This Item

EC Number:
UNSPSC Code:
12352204

$570.00


Check Cart for Availability

recombinant

expressed in E. coli

Quality Level

description

of Pseudomonas fluorescens from E. coli

form

suspension

specific activity

80 U/mg (Approximately 80 U/mg protein at +25°C with galactose as the substrate.)
~80 units/mg protein (At 25 °C with galactose as the substrate.)

packaging

pkg of 50 U

manufacturer/tradename

Roche

shipped in

wet ice

storage temp.

2-8°C

Related Categories

General description

D-galactose:NAD+ 1-oxidoreductase

Application

β-Galactose Dehydrogenase S has been used in the colorimetric microassay method to determine the level of galactose and galactose-1-phosphate in blood.[1]

Quality

Contaminants: <0.01% ADH, <0.01% ß-galactosidase, <0.1% LDH, <0.05% NADH oxidase

Physical form

Suspension in 3.2 M ammonium sulfate solution, pH approximately 6

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

S Bandyopadhyay et al.
Human reproduction (Oxford, England), 18(10), 2031-2038 (2003-09-26)
The pathophysiological mechanisms underlying premature ovarian failure (POF) are largely unknown. Our objective was to develop a working animal model to explore the pathogenesis of POF. Since galactosaemic women eventually develop POF, we evaluated the potential of experimental galactose toxicity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service