Skip to Content
Merck
All Photos(1)

Key Documents

10109223001

Roche

RNA from yeast

dry powder, pkg of 100 g

Synonym(s):

RNA from yeast, RNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106311
Pricing and availability is not currently available.

biological source

yeast

Quality Level

form

dry powder

packaging

pkg of 100 g

manufacturer/tradename

Roche

color

yellowish to brown

solubility

water: soluble

storage temp.

room temp

Related Categories

General description

RNA from yeast is the total RNA obtained from Candida utilis.

Application

RNA from yeast has been used:
  • as a carrier RNA for in situ hybridization[1]
  • for RNA coimmunoprecipitation[2]
RNA from yeast has been used:
  • to investigate its effect on the formation of transactivation response element (TAR) DNA-binding protein 43 (TDP-43) low-complexity domain (LCD) droplets in phosphate buffer to determine the possible influence of RNA binding on the liquid–liquid phase separation (LLPS) and/or aggregation of TDP-43 LCD[3]
  • for biomass fractionation to help with separations to assess the fate of methylphosphonate (MPn) or bicarbonate radiotracers in cells[4]
  • in hybridization solution for in situ hybridization[5]

Preparation Note

Working solution: This mixture of total yeast RNA is resuspended with sterile, DEPC-treated double-distilled water or TE buffer to 10 mg/ml if used as carrier.

Analysis Note

Absorbance: RNA: 95%, Pi (phosphate): 0.3%

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donya Pakravan et al.
Biomolecules, 11(4) (2021-05-01)
Aggregates of TAR DNA-binding protein (TDP-43) are a hallmark of several neurodegenerative disorders, including amyotrophic lateral sclerosis (ALS). Although TDP-43 aggregates are an undisputed pathological species at the end stage of these diseases, the molecular changes underlying the initiation of
Britta Jedamzik et al.
Cold Spring Harbor protocols, 2009(10), pdb-pdb (2010-02-12)
RNA coimmunoprecipitation (co-IP) experiments are an extension of protein co-IP experiments in which in vivo RNA-protein complexes are investigated. This protocol describes how to perform RNA co-IPs from C. elegans whole-worm extracts. In principle, a protein-specific antibody is used to
Simona Abbatemarco et al.
Journal of cell science, 134(22) (2021-10-19)
When exposed to stressful conditions, eukaryotic cells respond by inducing the formation of cytoplasmic ribonucleoprotein complexes called stress granules. Here, we use C. elegans to study two proteins that are important for stress granule assembly in human cells - PQN-59
Anja Werner et al.
iScience, 24(9), 103076-103076 (2021-09-30)
Systemic lupus erythematosus (SLE) is characterized by a loss of self-tolerance, systemic inflammation, and multi-organ damage. While a variety of therapeutic interventions are available, it has become clear that an early diagnosis and treatment may be key to achieve long
Karen Crawford et al.
Current biology : CB, 30(17), 3484-3490 (2020-08-01)
Seminal studies using squid as a model led to breakthroughs in neurobiology. The squid giant axon and synapse, for example, laid the foundation for our current understanding of the action potential [1], ionic gradients across cells [2], voltage-dependent ion channels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service