Key Documents
ZIXUVLPA1
Milli-Q
ech2o® UV lamp
Protects stored water against biofilm buildup in Milli-Q® IX & IQ 7003/05/10/15 storage tanks
Select a Size
1 KIT
$555.00
$555.00
Available to ship on11 April 2025Details
Request more information
Select a Size
Change View
1 KIT
$555.00
About This Item
UNSPSC Code:
41104200
NACRES:
NB.33
$555.00
Available to ship on11 April 2025Details
Request more information
Quality Level
packaging
pkg of 1 unit
compatibility
for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015
General description
ECH2O® mercury-free UV LED lamp is used to control bacterial growth and preserves water quality. This UV Lamp can be used with Milli-Q® IQ 7003/05/10/15 and Milli-Q® IX 7003/05/10/15 water purification systems.
Application
The ECH2O® lamp is used to limit the bacteria level and prevents biofilm formation in the storage tanks.
Legal Information
ECH2O is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Questions
1–7 of 7 Questions  
  1. What are the storage conditions for this product?
    1 answer
    1. Recommended storage conditions: temperatures from 5 to 30°C; humidity between 10 and 80%. The original sealed bag (if provided) must be kept intact before the first use to protect the purification element from foreign matter.
      Helpful?
  2. How can I know which consumables can be used with my system?
    1 answer
    1. To know which consumables can be used on your system, you can see in the user manual of your system or see all the consumables for each system listed on https://www.sigmaaldrich.com/products/water-purification/milli-q-consumables.
      Helpful?
  3. What steps are you taking towards improved sustainability?
    1 answer
    1. Sustainability is at the heart of our innovations. From reducing water and energy consumption, to minimizing plastic and chemical waste, our dedicated water system engineers are constantly working on new ways to reduce the ecological footprint of our products to better support the sustainability goals we're all striving towards. For more information, we invite you to review the following brochure and infographic which highlight our efforts and achievements: https://www.sigmaaldrich.com/life-science/milliq#sus. Do not hesitate to contact your local representative or sales contact (https://www.sigmaaldrich.com/collections/offices) for further information.
      Helpful?
  4. Can I arrange to have my consumables automatically ordered and delivered once a year?
    1 answer
    1. It's possible to place only one consumable order per year with a specific contract called "Consumable Supply Agreement (CSA)" (https://www.sigmaaldrich.com/US/en/products/water-purification/milli-q-consumables#csa). For further information, contact your local representative or sales contact (https://www.sigmaaldrich.com/US/en/collections/offices).
      Helpful?
  5. How frequently do the various consumables need to be changed?
    1 answer
    1. The frequency that consumables must be changed depends on the system's usage (treated water volume) and on the consumables themselves (time of usage). In order to ensure the quality of water and your results, the consumables should be replaced when the system alerts you.
      Helpful?
  6. Where can I find data sheets, Certificates of Origin, Certificates of Analysis, and Certificates of Quality?
    1 answer
    1. Documents and certificates are available in the "Documentation" section on this page. Do not hesitate to contact us if you require further information.
      Helpful?
  7. Can I change the UV lamp myself?
    1 answer
    1. No. With the exception of our SMART system range, a field service engineer must change the UV lamp as this process can be dangerous for customers and requires special protective equipment.
      Helpful?