Skip to Content
Merck
All Photos(2)

Documents

YY2209064

Millipore

Support Screen 90 mm, stainless steel

Accessories for filter holders for sample preparation.

Synonym(s):

Stainless steel support

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24

material

stainless steel

Quality Level

diam.

90 mm

compatibility

for use with Filter Holder 90 mm, stainless steel

shipped in

ambient

General description

Do your waste samples contain volatile substances, such as isopropanol, acetone or xylene? Then use our ZHE (Zero Head Space Extractor) Hazardous Waste Filtration system, which is approved by the US Environmental Protection Agency (EPA) as an apparatus for Toxicity Characteristic Leaching Procedure (TCLP) measurement.



The system cannot pressurize unless completely assembled; this eliminates accidentally “shooting the piston” out of the unit and assures safe operation. Choose both manual and automatic venting modes.



Features & Benefits:

•Allows for pressurization of filter without introducing air, due to movable piston design

•Prevents loss of volatiles

•Manual and automatic venting modes

•System cannot be pressurized unless completely assembled, thereby preventing "shooting the piston" out of the unit accidentally



Applications:

US EPA Extraction Procedure (EP) Toxicity Test and Toxicity Characteristic Leaching Procedure



Hazardous Waste System

YT30090HW Millipore ZHE Hazardous Waste System



Replacement Parts

XX2702512 2-way Valve, 1/8 in. NPTM, stainless steel

XX6700010 Cylinder O-rings 3

XX6700024 Vent/Relief Valve, 9 bar

XX6700104 Street Elbow, 1/4 in. NPTF to M

YT3009001 Piston with O-rings, wiper seal

YT3009002 Fitting Luer 1/8 in. NPTF

YT3009003 Piston O-rings, wiper seal

YT3009013 Quick-disconnect Female Swagelok

YT3009014 Quick-disconnect Male Swagelok

YY2209064 Support Screen 90 mm, stainless steel

YY2209068 O-ring, 2-235, 90 mm, fluoroelastomer

YY2214257 Hand Knob



Accessories

XX6700P05 Dispensing Pressure Vessel, 5 L

YT3009015 ZHE Tight Syringe, 50ml

YT3009016 ZHE Tedlar Bag, 1 liter



Connector Kit Components

XX3002567 Adapter, 1/8 in. NPTF to M Luer-Lok, chrome plated

XX6700024 Vent/Relief Valve, 9 bar

XX6700030 Quick-release Nipple & Coupling, 1/4 in. NPTM

XX6700125 Hex Nipple, 1/4 in. NPTM 51 mm., stainless steel

XX6702507 Tube Adapter, 1/4 in. NPTM to 1/4 in. Tube

XX6702508 Tubing, 6 mm O.D. x 3 m, polypropylene

YT3009013 Quick-disconnect Female Swagelok

YY1301015 Pressure Gauge, 0–7 bar

YY2029348 Ball Valve, 1/4 in. NPTF, stainless steel

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service