Merck
All Photos(4)
Key Documents
TZNFVSP20
Millipore
Steritest® Sampling Needle-Free Valve
sterile; ethylene oxide treated
Select a Size
20
$128.00
$128.00
Check Cart for Availability
Select a Size
Change View
20
$128.00
About This Item
UNSPSC Code:
23151818
NACRES:
JA.12
$128.00
Check Cart for Availability
Agency
USP 88
Quality Level
sterility
sterile; ethylene oxide treated
packaging
box of 10 units (2 valves per package)
application(s)
pharmaceutical
sterility testing
General description
Steritest® Sampling Needle-Free Valve is designed as an accessory which is installed on Steritest® Neo Devices. The valve has a male luer slip and a female luer lock with a septum. The valve can be activated by a male luer without a needle.
The unique design allows aseptic installation (inside an isolator) post media filling step during sterility testing. The installation is recommended when a sampling of the media is planned.
Post installation, the media inside the canisters can be accessed through a luer syringe. Sampled media from Steritest® canisters can be further used for:
  • Turbidity detected during or after incubation
  • Use of rapid detection method (e.g: ATP measurement)
Application
Steritest® Sampling Needle-Free Valve is used to sample an aliquot of media from the canisters during or after incubation during sterility testing.
Legal Information
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany